Proteintech
Manufacturer Code:664061IG150UL
Catalog # 50-173-6834
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E2F10 |
Immunogen | CD80/B7-1 Fusion Protein Ag5615 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activation B7 1 antigen B7 B7 1 B7-1 BB1 CD28LG CD28LG1 CD80 CD80 molecule CTLA 4 counter receptor B7.1 LAB7; Activation B7-1 antigen; B-lymphocyte activation antigen B7; B7; B7 protein; BB1; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; T-cell costimulatory molecule; T-lymphocyte activation antigen CD80
Gene Aliases: B7; B7-1; B7.1; B71; BB1; Cd28l; CD28LG; CD28LG1; CD80; LAB7; Ly-53; Ly53; MIC17; TSA1
UniProt ID: (Human) P33681, (Mouse) Q00609
Entrez Gene ID: (Human) 941, (Pig) 397161, (Mouse) 12519
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.