Invitrogen
Manufacturer Code:160801050
Catalog # 16-080-1050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | 16-10A1 |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: Activation B7-1 antigen; Activation B7-1 antigen B7 B7 protein B7.1 B71 B7-1 B7-1 protein BB1 B-lymphocyte activation antigen B7 Cd28l CD28LG CD28LG1 CD80 Cd80 a rat homolog of the human CD28/CTLA - 4 ligand (B7-1) CD80 anitgen CD80 antigen CD80 antigen (CD28 antigen ligand 1 B7-1 antigen) Cd80 molecule CD80 protein precursor costimulatory factor CD80 costimulatory molecule variant IgV-CD80 CTLA-4 counter-receptor B7.1 LAB7 Ly53 Ly-53 MIC17 sCD 80 sCD80 soluble CD 80 soluble CD80 T-cell costimulatory molecule T-cell co-stimulatory protein B7-1 T-lymphocyte activation antigen CD80 TS/A-1 TSA1; B7; B7 protein; B7-1 protein; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); T-cell co-stimulatory protein B7-1; T-cell costimulatory molecule; T-lymphocyte activation antigen CD80
Gene Aliases: B7; B7-1; B71; Cd28l; Cd80; Ly-53; Ly53; MIC17; TSA1
UniProt ID: (Mouse) Q00609
Entrez Gene ID: (Pig) 397161, (Dog) 403765, (Mouse) 12519
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.