OriGene
Manufacturer Code:TA501669
Catalog # 50-167-2793
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7H3 |
Immunogen | Full length human recombit protein of human CD80 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activation B7-1 antigen; B-lymphocyte activation antigen B7; B7; B7 B7-1 B7.1 BB1 CD28LG CD28LG1 LAB7; BB1; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; T-lymphocyte activation antigen CD80
Gene Aliases: B7; B7-1; B7.1; BB1; CD28LG; CD28LG1; CD80; LAB7
UniProt ID: (Human) P33681
Entrez Gene ID: (Human) 941
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.