Invitrogen
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | 5 µL/1x10^6 cells |
Host / Isotype | Mouse / IgG1 |
Class | Monoclonal |
Type | Antibody |
Clone | MEM-233 |
Immunogen | Extracellular domain of human CD80 fused to human IgG1(Fc) |
Conjugate |
Alexa Fluor™ 647
|
Form | Liquid |
Concentration | 0.12 mg/mL |
Storage conditions | 4° C, store in dark |
RRID | AB_2633302 |
CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28, and of an inhibitory receptor CTLA-4 (CD152). Both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, and both molecules can also substitute for each other in this process. CD80 is rapidly induced on the surface of in vitro activated B cells, with Epstein Barr Virus (EBV) transformed B cell lines, with Burkitts lymphoma cell lines, with freshly isolated follicular B lymphoma cells, T cells, and monocytes. The B-lymphocyte activation antigen B7-1 provides regulatory signals for T lymphocytes as a consequence of binding to the CD28 and CTLA4 ligands of T cells. Diseases associated with CD80 dysfunction include gallbladder squamous cell carcinoma and myocarditis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.