Novus Biologicals
Manufacturer Code:NBP149045
Catalog # NBP149045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 53-6.7 |
Immunogen | Mouse thymus or spleen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: 53-6.7 53-6.7 CD8 53-6.7 CD8 Alpha 53-6.7 CD8 Aplha 53-6.7 Clone CD8 CD8a molecule Cytotoxic T cell marker Cytotoxic T lymphocyte marker LEU2 p32 T cell marker; CD8 antigen, alpha polypeptide (p32); CD8a; Leu2 T-lymphocyte antigen; OKT8 T-cell antigen; T cell co-receptor; T-cell antigen Leu2; T-cell surface glycoprotein CD8 alpha chain; T-lymphocyte differentiation antigen T8/Leu-2; T8 T-cell antigen
Gene Aliases: CD8; CD8A; Leu2; MAL; p32
UniProt ID: (Human) P01732
Entrez Gene ID: (Human) 925
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.