Novus Biologicals
Manufacturer Code:NB1001985
Catalog # NB1001985
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | PIN.1 |
Immunogen | Peptide corresponding to residues 12-27 of CD74 human invariant chain (short form). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD74; CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated); CD74 antigen CD74 antigen (invariant polypeptide of major histocompatibility complex classII antigen-associated) CD74 molecule major histocompatibility complex class II invariant chain DHLAGgamma chain of class II antigens HLA class II histocompatibility antigen gamma chain HLADG HLA-DR antigens-associated invariant chain HLA-DR-gamma Ia antigen-associated invariant chain Ia-associated invariant chain Ia-GAMMA Ii MHC HLA-DR gamma chain p33; CD74 molecule, major histocompatibility complex, class II invariant chain; Class-II-associated invariant chain peptide; CLIP; gamma chain of class II antigens; HLA class II histocompatibility antigen gamma chain; HLA-DR antigens-associated invariant chain; HLA-DR-gamma; Ia antigen-associated invariant chain; Ia-associated invariant chain; Ii; MHC HLA-DR gamma chain; p33
Gene Aliases: CD74; DHLAG; HLADG; Ia-GAMMA; II
UniProt ID: (Human) P04233
Entrez Gene ID: (Human) 972
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.