Invitrogen
Manufacturer Code:160731025
Catalog # 16-073-1025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Control (Ctrl) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | eBioTY/11.8 (TY/11.8) |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: 2210401F01Rik 5 nucleotidase ecto 5' nucleotidase ecto 5'-NT 5'-nucleotidase 5'-nucleotidase ecto 5'-nucleotidase ecto (CD73) AI447961 CALJA CD73 E5NT Ecto-5'-nucleotidase eN eNT NT Nt5 Nt5e Nte Purine 5-Prime-Nucleotidase; 5'-NT; 5'-nucleotidase; CD73; Ecto-5'-nucleotidase
Gene Aliases: 2210401F01Rik; AI447961; CD73; eNT; NT; Nt5; Nt5e; Nte
UniProt ID: (Mouse) Q61503
Entrez Gene ID: (Mouse) 23959
Molecular Function: phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.