Invitrogen
Manufacturer Code:17073941
Catalog # 50-150-22
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AD2 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 5'-NT; 5'-nucleotidase; 5'-nucleotidase, ecto (CD73); CD73; CD73 E5NT NT NT5 NTE eN eNT 5&prime-NT 5&prime-nucleotidase Purine 5-Prime-Nucleotidase ecto-5&prime-nucleotidase; Ecto-5'-nucleotidase; Purine 5-Prime-Nucleotidase
Gene Aliases: CALJA; CD73; E5NT; eN; eNT; NT; NT5; NT5E; NTE
UniProt ID: (Human) P21589
Entrez Gene ID: (Human) 4907
Molecular Function: phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.