Invitrogen
Manufacturer Code:PA527336
Catalog # PIPA527336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 211 and 538 of Human CD73 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-NT; 5'-nucleotidase; 5'-nucleotidase, ecto (CD73); CD73; CD73 E5NT NT NT5 NTE eN eNT 5&prime-NT 5&prime-nucleotidase Purine 5-Prime-Nucleotidase ecto-5&prime-nucleotidase; Ecto-5'-nucleotidase; Purine 5-Prime-Nucleotidase
Gene Aliases: 2210401F01Rik; AI447961; CALJA; CD73; E5NT; eN; eNT; NT; NT5; NT5E; NTE
UniProt ID: (Human) P21589, (Mouse) Q61503
Entrez Gene ID: (Human) 4907, (Mouse) 23959
Molecular Function:
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.