Invitrogen
Manufacturer Code:PA511871
Catalog # PIPA511871
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 520-550 amino acids from the C-terminal region of human CD73 (NT5E) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-NT; 5'-nucleotidase; 5'-nucleotidase, ecto (CD73); CD73; CD73 E5NT NT NT5 NTE eN eNT 5&prime-NT 5&prime-nucleotidase Purine 5-Prime-Nucleotidase ecto-5&prime-nucleotidase; Ecto-5'-nucleotidase; Purine 5-Prime-Nucleotidase
Gene Aliases: 2210401F01Rik; AI447961; CALJA; CD73; E5NT; eN; eNT; NT; NT5; NT5E; NTE
UniProt ID: (Human) P21589, (Mouse) Q61503
Entrez Gene ID: (Human) 4907, (Mouse) 23959
Molecular Function:
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.