Invitrogen
Manufacturer Code:14071982
Catalog # 50-126-98
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OKT9 (OKT-9) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD71; CD71 T9 TFR TFR1 TR TRFR p90 transferrin receptor protein 1; p90; sTfR; T9; TR; Transferrin receptor protein 1; Transferrin receptor protein 1, serum form
Gene Aliases: CD71; IMD46; p90; T9; TFR; TFR1; TFRC; TR; TRFR
UniProt ID: (Human) P02786
Entrez Gene ID: (Human) 7037
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.