Invitrogen
Manufacturer Code:14066195
Catalog # 14-066-195
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: antigen CD66 ATP-dependent taurocolate-carrier protein bb-1 BGP BGP1 BGP-1 BGPI BGPR Biliary glycoprotein 1 Biliary glycoprotein D bone gamma-carboxyglutamic acid (Gla) protein carcinoembryonic antigen 1 carcinoembryonic antigen 7 carcinoembryonic antigen related cell adhesion molecule 1 carcinoembryonic antigen-related cell adhesion molecule 1 carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein) Cc1 C-CAM C-CAM 105 Ccam1 CD66a CD66a antigen Cea1 Cea-1 Cea7 Cea-7 CEACAM1 CEA-related cell adhesion molecule 1 CEA-related cell adhesion molecule 1 (bone gamma-carboxyglutamic acid (Gla) protein) (osteocalcin) cell-CAM 105 Ecto-ATPase GP110 hepatitis virus (MHV-4) susceptibility hepatitis virus receptor Hv2 Hv-2 mCEA1 Mhv-1 MHVR MHV-R MHVR1 mmCGM1 mmCGM1a mmCGM2 Murine hepatitis virus receptor osteocalcin pp120 pp120/ecto-ATPase; BGP-1; Biliary glycoprotein 1; Biliary glycoprotein D; carcinoembryonic antigen 1; carcinoembryonic antigen 7; Carcinoembryonic antigen-related cell adhesion molecule 1; CD66a; CEA-related cell adhesion molecule 1; hepatitis virus (MHV-4) susceptibility; hepatitis virus receptor; MHV-R; MHVR1; Murine hepatitis virus receptor
Gene Aliases: bb-1; Bgp; Bgp1; C-CAM; Cc1; CD66a; Cea-1; Cea-7; Cea1; Cea7; Ceacam1; Hv-2; Hv2; mCEA1; Mhv-1; MHVR; MHVR1; mmCGM1; mmCGM1a; mmCGM2
UniProt ID: (Mouse) P31809
Entrez Gene ID: (Mouse) 26365
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
kinase
membrane-bound signaling molecule
protein kinase
receptor
signaling molecule
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.