Invitrogen
Manufacturer Code:25064941
Catalog # 25-064-941
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10.1 |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD64; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc gamma receptor Ib; Fc-gamma receptor I A1; Fc-gamma receptor I B2; Fc-gamma RI; Fc-gamma RIA; fc-gamma RIB; FcgammaRIa; FcRI; fcRIB; hFcgammaRIB; High affinity immunoglobulin gamma Fc receptor I; IgG Fc receptor I; igG Fc receptor IB
Gene Aliases: CD64; CD64A; CD64b; FCG1; FCGR1; FCGR1A; FCRI; IGFR1; IGFRB
UniProt ID: (Human) P12314
Entrez Gene ID: (Human) 2209, (Human) 2210
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.