Novus Biologicals
Manufacturer Code:NBP29276320UL
Catalog # NB027396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of Human FCGR1B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD64b Fc fragment of IgG high affinity Ib receptor (CD64) IGFRB; Fc fragment of IgG, high affinity Ib, receptor (CD64); Fc gamma receptor Ib; Fc-gamma receptor I B2; fc-gamma RIB; fcRIB; hFcgammaRIB; igG Fc receptor IB
Gene Aliases: CD64b; IGFRB
Entrez Gene ID: (Human) 2210
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.