Invitrogen
Manufacturer Code:MA530588
Catalog # PIMA530588
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B3B1C9 |
Immunogen | Recombit Human CD64 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD64; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRIa; FcRI; High affinity immunoglobulin gamma Fc receptor I; IgG Fc receptor I; RP11-196G18.2 CD64 CD64A FCRI FLJ18345 IGFR1 Fc fragment of IgG high affinity Ia receptor for CD64 Fc gamma receptor Fc-gamma RI Fc-gamma receptor 1
Gene Aliases: CD64; CD64A; FCG1; FCGR1; FCGR1A; FCRI; IGFR1
UniProt ID: (Human) P12314
Entrez Gene ID: (Human) 2209
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.