Invitrogen
Manufacturer Code:MA523993
Catalog # PIMA523993
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 290322 |
Immunogen | NS0-derived recombit mouse Fc gamma RI extracellular domain Glu25-Pro297 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD64; CD64 FCGR1A FCRI CD64 antigen Fc fragment of IgG high affinity Ia receptor (CD64) FCG1 Fc-gamma receptor I B2 Fc-gamma RI Fc-gamma RIA FcgammaRIa FCGR1 FcRI FCRI FLJ18345 high affinity Ia receptor for (CD64) high affinity immunoglobulin gamma Fc receptor I IgG Fc receptor I; Fc-gamma RI; FcRI; High affinity immunoglobulin gamma Fc receptor I; IgG Fc receptor I; IgG high affinity Fc receptor
Gene Aliases: AI323638; AV092959; CD64; Fcg1; FcgammaRI; Fcgr1; IGGHAFC
UniProt ID: (Mouse) P26151
Entrez Gene ID: (Mouse) 14129
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.