Invitrogen
Manufacturer Code:MA530590
Catalog # PIMA530590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 15 |
Immunogen | Recombit Rat CD64/FCGR1A protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc receptor, IgG, high affinity I; high affinity IgG Fc receptor, subunit beta; high affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; RP11-196G18.2 CD64 CD64A FCRI FLJ18345 IGFR1 Fc fragment of IgG high affinity Ia receptor for CD64 Fc gamma receptor Fc-gamma RI Fc-gamma receptor 1
Gene Aliases: FcgammaRI; Fcgr1; Fcgr1b
Entrez Gene ID: (Rat) 295279
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.