Invitrogen
Manufacturer Code:48064941
Catalog # 48-064-941
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10.1 |
Conjugate | eFluor 450 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AI323638 AV092959 CD64 CD64 antigen CD64A Fc fragment of IgG high affinity Ib receptor Fc fragment of IgG receptor Ia Fc fragment of IgG high affinity Ia receptor (CD64) Fc fragment of IgG high affinity Ia receptor for (CD64) Fc fragment of IgG high affinity Ib receptor for (CD64) Fc gamma receptor Fc gamma receptor Ia Fc receptor IgG high affinity I FCG1 Fc-gamma receptor 1 Fc-gamma receptor I A1 fc-gamma RI Fc-gamma RIA FcgammaRI FcgammaRIa Fcgr1 FCGR1A Fcgr1b FcgRI FCRI FLJ18345 high affinity IgG Fc receptor subunit beta high affinity immunoglobulin gamma Fc receptor I high-affinity immunoglobulin gamma Fc receptor I IGFR1 IgG Fc receptor I IgG high affinity Fc receptor IGGHAFC RP11-196G18.2; CD64; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRIa; FcRI; High affinity immunoglobulin gamma Fc receptor I; IgG Fc receptor I
Gene Aliases: CD64; CD64A; FCG1; FCGR1; FCGR1A; FCRI; IGFR1
UniProt ID: (Human) P12314
Entrez Gene ID: (Human) 2209
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.