Invitrogen
Manufacturer Code:PA581386
Catalog # PIPA581386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Mouse FCGR1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD64; Fc-gamma RI; FcRI; High affinity immunoglobulin gamma Fc receptor I; IgG Fc receptor I; IgG high affinity Fc receptor; RP11-196G18.2 CD64 CD64A FCRI FLJ18345 IGFR1 Fc fragment of IgG high affinity Ia receptor for CD64 Fc gamma receptor Fc-gamma RI Fc-gamma receptor 1
Gene Aliases: AI323638; AV092959; CD64; Fcg1; FcgammaRI; Fcgr1; IGGHAFC
UniProt ID: (Mouse) P26151
Entrez Gene ID: (Mouse) 14129
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.