Invitrogen
Manufacturer Code:PA5114987
Catalog # PIPA5114987
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CD64 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI323638 AV092959 CD64 CD64 antigen CD64A Fc fragment of IgG high affinity Ib receptor Fc fragment of IgG receptor Ia Fc fragment of IgG high affinity Ia receptor (CD64) Fc fragment of IgG high affinity Ia receptor for (CD64) Fc fragment of IgG high affinity Ib receptor for (CD64) Fc gamma receptor Fc gamma receptor Ia Fc receptor IgG high affinity I FCG1 Fc-gamma receptor 1 Fc-gamma receptor I A1 fc-gamma RI Fc-gamma RIA FcgammaRI FcgammaRIa Fcgr1 FCGR1A Fcgr1b FcgRI FCRI FLJ18345 high affinity IgG Fc receptor subunit beta high affinity immunoglobulin gamma Fc receptor I high-affinity immunoglobulin gamma Fc receptor I IGFR1 IgG Fc receptor I IgG high affinity Fc receptor IGGHAFC RP11-196G18.2; CD64; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc receptor, IgG, high affinity I; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRIa; FcRI; high affinity IgG Fc receptor, subunit beta; High affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; IgG Fc receptor I
Gene Aliases: CD64; CD64A; FCG1; FcgammaRI; FCGR1; FCGR1A; Fcgr1b; FCRI; IGFR1
UniProt ID: (Human) P12314
Entrez Gene ID: (Human) 2209, (Rat) 295279
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.