Invitrogen
Manufacturer Code:14061982
Catalog # 50-126-68
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | VI-PL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: antigen CD61; BDPLT2 CD61 GP3A GPIIIa GT integrin beta-3 platelet glycoprotein IIIa platelet membrane glycoprotein IIIa ITGB3; CD61; glycoprotein GPIIIa; GPIIIa; integrin beta 3; Integrin beta-3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); Platelet membrane glycoprotein IIIa
Gene Aliases: BDPLT16; BDPLT2; CD61; EGK_08849; GP3A; GPIIIA; GT; ITGB3
UniProt ID: (Human) P05106
Entrez Gene ID: (Human) 3690, (Cynomolgus monkey) 102117791, (Dog) 403788, (Rhesus monkey) 717379
Molecular Function: cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.