Invitrogen
Manufacturer Code:13061181
Catalog # 50-116-02
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | 2C9.G3 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: BDPLT2 CD61 GP3A GPIIIa GT integrin beta-3 platelet glycoprotein IIIa platelet membrane glycoprotein IIIa ITGB3; CD61; GPIIIa; integrin beta 3 (Cd61); Integrin beta-3; platelet gpIIIa; Platelet membrane glycoprotein IIIa
Gene Aliases: CD61; GP3A; INGRB3; Itgb3
UniProt ID: (Rat) Q8R2H2, (Mouse) O54890
Entrez Gene ID: (Rat) 29302, (Mouse) 16416
Molecular Function:
cell adhesion molecule
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.