Invitrogen
Manufacturer Code:MA530256
Catalog # PIMA530256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 45 |
Immunogen | Recombit Mouse CD59a/Protectin Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CD59; CD59 antigen; CD59 CD59 molecule complement regulatory protein 1F5 EJ16 EJ30 EL32 G344 MIN1 MIN2 MIN3 MIRL HRF20 MACIF MEM43 MIC11 MSK21 16.3A5 HRF-20 MAC-IP p18-20; CD59A glycoprotein; MAC-inhibitory protein; MAC-IP; MACIF; Membrane attack complex inhibition factor; Protectin
Gene Aliases: AA987121; Cd59; Cd59a; protectin
UniProt ID: (Mouse) O55186
Entrez Gene ID: (Mouse) 12509
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.