Invitrogen
Manufacturer Code:PA578994
Catalog # PIPA578994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse CD59 recombit protein (Position: L24-S96). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cb59b molecule; CD59; CD59 antigen; CD59 glycoprotein; CD59A glycoprotein; CD59A glycoprotein MAC-inhibitory protein MAC-IP Membrane attack complex inhibition factor MACIF Protectin CD59 Cd59a Cd59; CD59b moleucle, complement regulatory protein; MAC-inhibitory protein; MAC-IP; MACIF; Membrane attack complex inhibition factor; Protectin
Gene Aliases: AA987121; Cd59; Cd59a; Cd59b; MAC-IP; MACIF; MACIP; protectin
UniProt ID: (Rat) P27274, (Mouse) O55186
Entrez Gene ID: (Rat) 25407, (Mouse) 12509
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.