Novus Biologicals
Manufacturer Code:NBP28926950UL
Catalog # NB020023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 28 |
Immunogen | Recombinant Human CD55/DAF protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CD55; CD55 antigen; CD55 antigen CD55 molecule decay accelerating factor for complement (Cromer blood group) CRdecay accelerating factor for complement (CD55 Cromer blood group system) CROMDAFcomplement decay-accelerating factor decay accelerating factor for complement TC; CD55 molecule, decay accelerating factor for complement (Cromer blood group); Complement decay-accelerating factor
Gene Aliases: CD55; CR; CROM; DAF; TC
UniProt ID: (Human) Q14UF6
Entrez Gene ID: (Human) 1604
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.