R&D Systems
Manufacturer Code:AF2009
Catalog # AF2009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human CD55/DAF Asp35-Ser353 Accession # P08174 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD55; CD55 antigen; CD55 CD55 antigen CD55 molecule decay accelerating factor for complement (Cromer blood group) CR CRdecay accelerating factor for complement (CD55 Cromer blood group system) CROMDAFcomplement decay-accelerating factor DAF decay accelerating factor for complement TC; CD55 molecule, decay accelerating factor for complement (Cromer blood group); Complement decay-accelerating factor
Gene Aliases: CD55; CR; CROM; DAF; TC
UniProt ID: (Human) Q14UF6
Entrez Gene ID: (Human) 1604
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.