Invitrogen
Manufacturer Code:PA580745
Catalog # PIPA580745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the C-terminus of the human ITGAV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antigen identified by monoclonal antibody L230; CD51; CD51 MSK8 VNRA antigen identified by monoclonal antibody L230 integrin alpha-V vitronectin receptor subunit alpha ITGAV; integrin alpha V; Integrin alpha-V; Integrin alpha-V heavy chain; Integrin alpha-V light chain; integrin alphaVbeta3; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); Vitronectin receptor; Vitronectin receptor subunit alpha
Gene Aliases: CD51; ITGAV; MSK8; VNRA; VTNR
UniProt ID: (Human) P06756
Entrez Gene ID: (Human) 3685
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.