Novus Biologicals
Manufacturer Code:NBP221758
Catalog # NBP221758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SP19 |
Immunogen | Synthetic peptide from the intracellular region of human CD5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CD5; CD5 antigen (p56-62); CD5 antigen CD5 antigen (p56-62) CD5 molecule LEU1T-cell surface glycoprotein CD5 Lymphocyte antigen T1/Leu-1 T1; Lymphocyte antigen T1/Leu-1; T-cell surface glycoprotein CD5
Gene Aliases: CD5; LEU1; T1
UniProt ID: (Human) P06127
Entrez Gene ID: (Human) 921
Molecular Function: hydrolase oxidase oxidoreductase protease receptor serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.