Invitrogen
Manufacturer Code:160491025
Catalog # 16-049-1025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | HMa2 |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha 2 subunit of VLA-2 receptor BDPLT9 BR CD49 antigen-like family member B CD49B Collagen receptor DX5 GPIa HPA-5 human platelet alloantigen system 5 integrin alpha 2 integrin alpha-2 integrin subunit alpha 2 integrin alpha 2 integrin alpha 2 (CD49B alpha 2 subunit of VLA-2 receptor) ITGA2 platelet antigen Br platelet glycoprotein GPIa platelet membrane glycoprotein Ia very late activation protein 2 receptor alpha-2 subunit VLA-2 VLA2 receptor VLA-2 receptor alpha 2 subunit VLA-2 subunit alpha VLAA2; CD49 antigen-like family member B; CD49b; Collagen receptor; GPIa; Integrin alpha-2; Platelet membrane glycoprotein Ia; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha
Gene Aliases: CD49B; DX5; Itga2
UniProt ID: (Mouse) Q62469
Entrez Gene ID: (Mouse) 16398
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.