Invitrogen
Manufacturer Code:47597182
Catalog # 50-112-9105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | DX5 |
Conjugate | APC-eFluor 780 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: BR CD49B GPIa VLA-2 VLAA2 ITGA2; CD49 antigen-like family member B; CD49b; Collagen receptor; GPIa; Integrin alpha-2; Platelet membrane glycoprotein Ia; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha
Gene Aliases: CD49B; DX5; Itga2
UniProt ID: (Mouse) Q62469
Entrez Gene ID: (Mouse) 16398
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.