Invitrogen
Manufacturer Code:46949042
Catalog # 50-112-3953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TS2/7 |
Conjugate | PerCP-eFluor 710 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD49 antigen-like family member A; CD49a; integrin alpha 1; Integrin alpha-1; integrin, alpha 1; Laminin and collagen receptor; very late activation protein 1; VLA-1
Gene Aliases: CD49a; ITGA1; VLA1
UniProt ID: (Human) P56199
Entrez Gene ID: (Human) 3672
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.