Invitrogen
Manufacturer Code:PA579525
Catalog # PIPA579525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human Integrin alpha 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD49 antigen-like family member A; CD49a; integrin alpha 1; Integrin alpha-1; Integrin alpha-1 CD49 antigen-like family member A Laminin and collagen receptor VLA-1 CD49a ITGA1; integrin, alpha 1; Laminin and collagen receptor; very late activation protein 1; VLA-1
Gene Aliases: CD49a; ITGA1; VLA1
UniProt ID: (Human) P56199, (Rat) P18614
Entrez Gene ID: (Human) 3672, (Rat) 25118
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.