Invitrogen
Manufacturer Code:MA516538
Catalog # PIMA516538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Pig |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6D8/8 |
Immunogen | Porcine peripheral blood mononuclear cells. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred,Store at 4°C or -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AHUS2 MCP MGC26544 MIC10 TLX TRA2.10; CD46; CD46 complement regulatory protein; Membrane cofactor protein
Gene Aliases: CD46; MCP
UniProt ID: (Pig) O02839
Entrez Gene ID: (Pig) 396922
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.