Novus Biologicals
Manufacturer Code:NBP215811
Catalog # NBP215811
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CD45. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B220 CD45 antigen CD45R EC 3.1.3.48 L-CA LY5 protein tyrosine phosphatase receptor type C receptor-type tyrosine-protein phosphatase C T200 glycoprotein T200 leukocyte common antigen T200receptor type c polypeptide; CD45; CD45 antigen; L-CA; Leukocyte common antigen; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein; T200 leukocyte common antigen
Gene Aliases: B220; CD45; CD45R; GP180; L-CA; LCA; LY5; PTPRC; T200
UniProt ID: (Human) P08575
Entrez Gene ID: (Human) 5788
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.