Proteintech
Manufacturer Code:201031AP150UL
Catalog # 50-560-916
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CD45 Fusion Protein Ag13712 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B220 CD45 CD45R CD45RA GP180 L CA LCA Leukocyte common antigen LY5 PTPRC RPTPC T200; CD45; CD45 antigen; L-CA; leucocyte common antigen; Leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein; T200 leukocyte common antigen
Gene Aliases: B220; CD45; CD45R; GP180; L-CA; LCA; loc; Ly-5; LY5; Lyt-4; PTPRC; RT7; T200
UniProt ID: (Human) P08575, (Mouse) Q78EF1, (Rat) P04157
Entrez Gene ID: (Human) 5788, (Mouse) 19264, (Rat) 24699
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.