Novus Biologicals
Manufacturer Code:NBP147386
Catalog # NBP147386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8E2F3 |
Immunogen | Purified recombinant fragment of human CD44 (628-699) expressed in E. coli. [Uniprot: P16070] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD44; CD44 antigen; CD44 antigen CD44 molecule (Indian blood group) CD44R CDw44 cell surface glycoprotein CD44 chondroitin sulfate proteoglycan 8 CSPG8 ECMR-III Epican Extracellular matrix receptor III GP90 lymphocyte homing/adhesion receptor HCELL hematopoietic cell E- and L-selectin ligand Heparan sulfate proteoglycan Hermes antigen homing function and Indian blood group system HUTCH-I Hyaluronate receptor IN LHR MC56 MDU2CD44 antigen (homing function and Indian blood group system) MDU3CDW44 MIC4MGC10468 Pgp1 PGP-1 PGP-I Phagocytic glycoprotein 1 Phagocytic glycoprotein I; CDw44; cell surface glycoprotein CD44; chondroitin sulfate proteoglycan 8; ECMR-III; Epican; Extracellular matrix receptor III; GP90 lymphocyte homing/adhesion receptor; hematopoietic cell E- and L-selectin ligand; Heparan sulfate proteoglycan; Hermes antigen; homing function and Indian blood group system; HUTCH-I; Hyaluronate receptor; PGP-1; PGP-I; Phagocytic glycoprotein 1; Phagocytic glycoprotein I; soluble CD44
Gene Aliases: CD44; CDW44; CSPG8; ECMR-III; HCELL; HUTCH-I; IN; LHR; MC56; MDU2; MDU3; MIC4; Pgp1
UniProt ID: (Human) P16070
Entrez Gene ID: (Human) 960
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.