Proteintech
Manufacturer Code:156751AP150UL
Catalog # 50-173-0394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CD44 Fusion Protein Ag8143 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD44; CD44 antigen; CD44 CD44 antigen CDW44 CSPG8 ECMR III Epican HCELL Heparan sulfate proteoglycan Hermes antigen HUTCH I Hyaluronate receptor IN LHR MC56 MDU2 MDU3 MIC4 MUTCH I PGP 1 PGP I Pgp1 Phagocytic glycoprotein 1 Phagocytic glycoprotein I; CDw44; cell surface glycoprotein CD44; Cell surface glycoprotein CD44 (hyaluronate binding protein); chondroitin sulfate proteoglycan 8; ECMR-III; Epican; Extracellular matrix receptor III; GP90 lymphocyte homing/adhesion receptor; hematopoietic cell E- and L-selectin ligand; Heparan sulfate proteoglycan; Hermes antigen; homing function and Indian blood group system; HUTCH-I; Hyaluronate receptor; Ly-24; Lymphocyte antigen 24; PGP-1; PGP-I; Phagocytic glycoprotein 1; Phagocytic glycoprotein I; soluble CD44
Gene Aliases: AU023126; AW121933; AW146109; CD44; CD44A; CDW44; CSPG8; ECMR-III; HCELL; HERMES; HUTCH-I; IN; LHR; Ly-24; MC56; MDU2; MDU3; METAA; MIC4; Pgp-1; Pgp1; RHAMM
UniProt ID: (Human) P16070, (Mouse) Q62061, (Rat) P26051
Entrez Gene ID: (Human) 960, (Mouse) 12505, (Rat) 25406
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.