Novus Biologicals
Manufacturer Code:NB10063790
Catalog # NB10063790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AK2 |
Immunogen | Made to Human CD42b |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Antigen CD42b-alpha; Antigen CD42b-alpha BSS CD42b CD42b antigen CD42b-alpha glycoprotein Ib (platelet) alpha polypeptide Glycoprotein Ibalpha GP1B GP-Ib alpha GPIbA GPIb-alpha MGC34595 platelet glycoprotein Ib alpha chain platelet membrane glycoprotein 1b-alpha subunit; CD42b; Glycocalicin; glycoprotein Ib (platelet), alpha polypeptide; GP-Ib alpha; mutant platelet membrane glycoprotein Ib-alpha; Platelet glycoprotein Ib alpha chain; platelet membrane glycoprotein 1b-alpha subunit; platelet membrane glycoprotein Ib-alpha
Gene Aliases: BDPLT1; BDPLT3; BSS; CD42B; CD42b-alpha; DBPLT3; GP1B; GP1BA; GPIbA; GPIbalpha; VWDP
UniProt ID: (Human) P07359
Entrez Gene ID: (Human) 2811
Molecular Function: cytokine extracellular matrix protein receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.