Novus Biologicals
Manufacturer Code:NBP266756
Catalog # NBP266756
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM11-34 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: CD154; CD154 CD154 antigen CD40 antigen ligand CD40 ligand CD40-L CD40LIGM gp39 hCD40L HIGM1 T-B cell-activating molecule T-BAM T-cell antigen Gp39 TNF-related activation protein TNFSF5IMD3 TRAPtumor necrosis factor (ligand) superfamily member 5 (hyper-IgM syndrome) tumor necrosis factor (ligand) superfamily member 5 Tumor necrosis factor ligand superfamily member 5; CD40 antigen ligand; CD40 ligand; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40-L; sCD40L; T-B cell-activating molecule; T-cell antigen Gp39; TNF-related activation protein; TRAP; tumor necrosis factor (ligand) superfamily member 5; Tumor necrosis factor ligand superfamily member 5
Gene Aliases: CD154; CD40L; CD40LG; gp39; hCD40L; HIGM1; IGM; IMD3; T-BAM; TNFSF5; TRAP
UniProt ID: (Human) P29965
Entrez Gene ID: (Human) 959
Molecular Function: cytokine signaling molecule tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.