Proteintech
Manufacturer Code:665021IG150UL
Catalog # 50-173-6925
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E6D10 |
Immunogen | CD40L/CD154 Fusion Protein Ag24941 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD154; CD154 CD40 L CD40 ligand CD40L CD40LG gp39 hCD40L HIGM1 IGM IMD3 T BAM T cell antigen Gp39 TNF related activation protein TNFSF5 TRAP; CD40 antigen ligand; CD40 ligand; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40-L; sCD40L; T-B cell-activating molecule; T-cell antigen Gp39; TNF-related activation protein; TRAP; tumor necrosis factor (ligand) superfamily member 5; Tumor necrosis factor ligand superfamily member 5
Gene Aliases: CD154; CD40L; CD40LG; gp39; hCD40L; HIGM1; IGM; IMD3; T-BAM; TNFSF5; TRAP
UniProt ID: (Human) P29965
Entrez Gene ID: (Human) 959
Molecular Function:
cytokine
signaling molecule
tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.