Novus Biologicals
Manufacturer Code:NBP1395560.025MG
Catalog # NB018510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | LOB7/6 |
Immunogen | Recognizes human CD40 cell surface antigen a 48kD glycoprotein expressed by B lymphocytes and weakly by some monocytes. CD40 is involved in the process of B cell selection in germinal centers and is vital in T cell-B cell interactions. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B cell surface antigen CD40; B cell surface antigen CD40 B-cell surface antigen CD40 Bp50B cell-associated molecule CD40 antigen CD40 molecule TNF receptor superfamily member 5 CD40 type II isoform CD40L receptor CDw40 MGC9013 nerve growth factor receptor-related B-lymphocyte activation molecule p50 TNFRSF5CD40 antigen (TNF receptor superfamily member 5) tumor necrosis factor receptor superfamily member 5 tumor necrosis factor receptor superfamily member 5; B cell-associated molecule; B-cell surface antigen CD40; Bp50; CD40; CD40 molecule, TNF receptor superfamily member 5; CD40L receptor; CDw40; Tumor necrosis factor receptor superfamily member 5
Gene Aliases: Bp50; CD40; CDW40; p50; TNFRSF5
UniProt ID: (Human) P25942
Entrez Gene ID: (Human) 958
Molecular Function: cytokine receptor receptor tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.