Invitrogen
Manufacturer Code:MA517009
Catalog # PIMA517009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Type | Antibody |
Clone | YTS191.1/KT15 |
Immunogen | T cell clone C6 |
Conjugate | FITC PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Antibody Cocktail
For Research Use Only
Protein Aliases: CD4; CD4 antigen p55 CD4 receptor T-cell surface antigen T4/Leu-3 T-cell surface glycoprotein CD4 CD8 Leu-2 MAL p32 CD8a CD8b CD8alpha CD8beta; CD8a; Lyt-2.1 lymphocyte differentiation antigen (AA at 100); T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein Lyt-2
Gene Aliases: BB154331; Cd4; Cd8a; L3T4; Ly-2; Ly-35; Ly-4; Ly-B; Lyt-2; Lyt2
UniProt ID: (Mouse) P06332, (Mouse) P01731
Entrez Gene ID: (Mouse) 12504, (Mouse) 12525
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.