Invitrogen
Manufacturer Code:MA516990
Catalog # PIMA516990
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Host / Isotype | Rat |
Type | Antibody |
Clone | YKIX302.9/YCATE55.9 |
Immunogen | Canine Concanavilin A activated T-cell blasts Canine CD8 alpha chimaeric human IgG1 Fc fusion protein. |
Conjugate | FITC PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Antibody Cocktail
For Research Use Only
Protein Aliases: CD4; CD4 antigen (p55); CD4 antigen p55 CD4 receptor T-cell surface antigen T4/Leu-3 T-cell surface glycoprotein CD4 CD8 Leu-2 MAL p32 CD8a CD8b CD8alpha CD8beta; CD8 antigen, alpha polypeptide (p32); CD8a; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD8 alpha chain
Gene Aliases: CD4; CD8A
UniProt ID: (Dog) P33705, (Dog) P33706
Entrez Gene ID: (Dog) 403931, (Dog) 403157, (Dog) 483076
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.