Invitrogen
Manufacturer Code:11504041
Catalog # 11-504-041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | YKIX302.9 |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD4; CD4 antigen (p55); CD4 CD4 antigen CD4 antigen (p55) CD4 antigen p55 Cd4 molecule CD4 receptor CD4 allele 1 cd4a CD4mut cell surface glycoprotein CD4 fCD4 L3T4 Leu-3 Ly-4 lymphocyte antigen CD4 lymphocyte antigen CD4 precursor p55 T-cell differentiation antigen L3T4 T-cell surface antigen T4/Leu-3 T-cell surface glycoprotein CD4 T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4) W3/25 W3/25 antigen; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4
Gene Aliases: CD4
UniProt ID: (Dog) P33705
Entrez Gene ID: (Dog) 403931
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.