OriGene
Manufacturer Code:TA500478
Catalog # 50-167-1611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9C1 |
Immunogen | Full length human recombit protein of human CD4 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD4; CD4 antigen; CD4 antigen (p55); CD4 receptor; CD4mut; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; W3/25 antigen
Gene Aliases: CD4; CD4mut; p55; W3/25
UniProt ID: (Dog) P33705, (Human) P01730, (Rat) P05540
Entrez Gene ID: (Dog) 403931, (Human) 920, (Rat) 24932
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.