Novus Biologicals
Manufacturer Code:NBP28982950UL
Catalog # NB021128
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 46 |
Immunogen | Recombinant Human CD39L4/ENTPD5 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CD39 antigen-like 4; CD39 antigen-like 4 CD39L4NTPDase 5 CD39-like 4 EC 3.6.1 EC 3.6.1.42 EC 3.6.1.6 ectonucleoside triphosphate diphosphohydrolase 5 ER-UDPase GDPase ENTPD5 Guanosine-diphosphatase ENTPD5 MGC163357 MGC163359 NTPDase-5 Nucleoside diphosphatase PCPH Pcph proto-oncogene protein proto-oncogene CPH UDPase ENTPD5 Uridine-diphosphatase ENTPD5; CD39-like 4; Ectonucleoside triphosphate diphosphohydrolase 5; ER-UDPase; GDPase ENTPD5; Guanosine-diphosphatase ENTPD5; NTPDase 5; Nucleoside diphosphatase; Pcph proto-oncogene protein; proto-oncogene CPH; UDPase ENTPD5; Uridine-diphosphatase ENTPD5
Gene Aliases: CD39L4; ENTPD5; NTPDase-5; PCPH
UniProt ID: (Human) O75356
Entrez Gene ID: (Human) 957
Molecular Function: hydrolase lyase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.