Novus Biologicals
Manufacturer Code:NBP132471
Catalog # NBP132471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ENTPD3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD39 antigen-like 3; CD39 antigen-like 3 CD39L3EC 3.6.1.5 CD39-like 3 EC 3.6.1 Ecto-apyrase 3 Ecto-ATP diphosphohydrolase 3 Ecto-ATPase 3 Ecto-ATPDase 3 ectonucleoside triphosphate diphosphohydrolase 3 FLJ93839 HB6NTPDase 3 NTPDase-3; Ecto-apyrase 3; Ecto-ATP diphosphohydrolase 3; ecto-ATPase 3; Ecto-ATPDase 3; Ectonucleoside triphosphate diphosphohydrolase 3; HB6; NTPDase 3
Gene Aliases: CD39L3; ENTPD3; HB6; NTPDase-3
UniProt ID: (Human) O75355
Entrez Gene ID: (Human) 956
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.