Novus Biologicals
Manufacturer Code:NBP132294
Catalog # NBP132294
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ENTPD6. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD39 antigen-like 2; CD39 antigen-like 2 CD39L2NTPDase-6 CD39-like 2 dJ738P15.3 DKFZp781G2277 DKFZp781K21102 EC 3.6.1.6 ectonucleoside triphosphate diphosphohydrolase 6 ectonucleoside triphosphate diphosphohydrolase 6 (putative function) ectonucleoside triphosphate diphosphohydrolase 6 (putative) FLJ36711 IL-6SAG IL6ST2 interleukin 6 signal transducer-2 NTPDase 6; Ectonucleoside triphosphate diphosphohydrolase 6; interleukin 6 signal transducer-2; NTPDase 6
Gene Aliases: CD39L2; dJ738P15.3; ENTPD6; IL-6SAG; IL6ST2; NTPDase-6
UniProt ID: (Human) O75354
Entrez Gene ID: (Human) 955
Molecular Function: hydrolase lyase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.