R&D Systems
Manufacturer Code:AF6087
Catalog # AF6087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human CD39L1/ENTPD2 Thr29-Asp460 Accession # Q9Y5L3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: CD39 antigen-like 1; CD39 antigen-like 1 CD39L1 CD39L1NTPDase 2 CD39-like-1 EC 3.6.1 EC 3.6.1.- EC 3.6.1.5 Ecto-ATP diphosphohydrolase 2 Ecto-ATPase Ecto-ATPase 2 Ecto-ATPDase 2 ectonucleoside triphosphate diphosphohydrolase 2 ENTPD2 NTPD2 NTPDase 2 NTPDase-2; Ecto-ATP diphosphohydrolase 2; ecto-ATPase 2; Ecto-ATPDase 2; Ectonucleoside triphosphate diphosphohydrolase 2; NTPDase 2
Gene Aliases: CD39L1; ENTPD2; NTPDase-2
UniProt ID: (Human) Q9Y5L3
Entrez Gene ID: (Human) 954
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.