Proteintech
Manufacturer Code:142111AP150UL
Catalog # 50-172-9285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CD39/ENTPD1 Fusion Protein Ag5429 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPDase; ATPDase CD39 Ecto apyrase Ecto ATP diphosphohydrolase 1 Ecto ATPase 1 Ecto ATPDase 1 ENTPD1 NTPDase 1; CD39; CD39 antigen; Ecto-apyrase; Ecto-ATP diphosphohydrolase 1; ecto-ATPase 1; Ecto-ATPDase 1; Ectonucleoside triphosphate diphosphohydrolase 1; Lymphoid cell activation antigen; NTPDase 1
Gene Aliases: 2610206B08Rik; AA408691; ATPDase; CD39; ENTPD1; NTPDase-1; SPG64
UniProt ID: (Human) P49961, (Mouse) P55772
Entrez Gene ID: (Human) 953, (Mouse) 12495, (Rat) 64519
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.